Learn More
DuPont™ Tychem™ 5000 Jacket
Catalog No. 17500130 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
2X-Large
3X-Large
4X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-500-130 Small
17-500-122 2X-Large
17-500-124 3X-Large
17-500-126 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 17-500-130 Supplier DuPont™ Supplier No. C3670TTNSM000600
Add to Cart
Add to Cart
This item is not returnable. View return policy
DuPont™ Tychem™ 5000 Jacket with Mandarin Collar, Elastic Wrists provides protection against a broad range of chemicals and situations involving liquid splash.
  • Ideal for applications that do not require the full body coverage of a coverall
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
  • Mandarin collar designed to fit tightly around hooded PAPR
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Typically worn with C3360T TN pants or sold as jacket/pant ensemble (C3750T TN)
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks
TRUSTED_SUSTAINABILITY
Specifications
Size Small
Gender Unisex
Material Tychem
Color Tan
Closure Type Double Hook and Loop Storm Flap
Arc Flash Rating None
Collar Type Mandarin Collar
Cuff Style Elastic
Seam Type Taped
Product Type Chemical-resistant Jacket