Learn More
DuPont™ Tychem™ 5000 Jacket
Catalog No. 17500123 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-500-123 2X-Large
17-500-125 3X-Large
17-500-127 4X-Large
17-500-128 Large
17-500-129 Medium
17-500-131 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-500-123 Supplier DuPont™ Supplier No. C3670TTN2X0006JF
Add to Cart
Add to Cart
This item is not returnable. View return policy
DuPont™ Tychem™ 5000 Jacket with Mandarin Collar, Jam Fit Cuff provides protection against a broad range of chemicals and situations involving liquid splash.
  • Ideal for applications that do not require the full body coverage of a coverall.
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Mandarin collar designed to fit tightly around hooded PAPR
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • Comes with a jam fit cuff and glove insert for user supplied gloves
  • Typically worn with C3360T TN pants or sold as jacket/pant ensemble (C3750T TN)
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
TRUSTED_SUSTAINABILITY
Specifications
Size 2X-Large
Gender Unisex
Material Tychem
Color Tan
Closure Type Zipper
Collar Type Mandarin Collar
Cuff Style Jam Fit
Seam Type Taped
Style Extends to Hip