Learn More
DuPont™ Tychem™ 6000 Apron
Catalog No. 18888836 Shop All DuPont Personal Protection Products
Change view
Click to view available options
TYCHEM™ 6000 Apron
Catalog No.
18-888-836 TYCHEM™ 6000 Apron
1 options
This item is not returnable. View return policy
Catalog No. 18-888-836 Supplier DuPont™ Supplier No. TYFPA30SGY00002500
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Tychem™ 6000 Aprons provide protection from chemical warfare agents and toxic industrial chemicals.
Tychem™ 6000 aprons are made of a proprietary barrier film laminated to a Tyvek™ substrate. Tychem™ 6000 fabrics are tested by a third party laboratory to assess its chemical permeation resistance.
  • Provides protection against over 180 chemical challenges for at least 30 minutes
  • Used by military personnel and first responders for chemical warfare situations
  • Available in low-visibility gray fabric for discretionary purpose
Certifications
Chemical Protective Clothing, Cat.III,Type Partial Body
TRUSTED_SUSTAINABILITY
Specifications
Type Apron
Size Universal
Material Tychem
Color Gray
Certifications/Compliance Chemical Protective Clothing, Cat.III,Type Partial Body