Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ 6000 Coverall with Attached Hood, Socks, Butyl Gloves

Catalog No. 18888793 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-888-793 X-Large
18-888-788 4X-Large
18-888-789 5X-Large
18-888-792 Small
18-888-786 2X-Large
18-888-787 3X-Large
18-888-790 Large
18-888-791 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 18-888-793 Supplier DuPont™ Supplier No. TF186TGYXL0006PI
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Coveralls provide protection from chemical warfare agents and toxic industrial chemicals.

  • Tychem™ 6000 coveralls are made of a proprietary barrier film laminated to a Tyvek™ substrate.
  • Tychem™ 6000 fabrics are tested by a third party laboratory to assess its chemical permeation resistance.
  • Provides protection against over 180 chemical challenges for at least 30 minutes
  • Used by military personnel and first responders for chemical warfare situations
  • Available in low-visibility gray fabric for discretionary purpose
TRUSTED_SUSTAINABILITY

Specifications

Color Gray
For Use With (Application) Military, Law Enforcement, Emergency Response
Hood Type Attached
Material Tychem
Size X-Large
Ankle Style Boot Overflaps
Cuff Style Attached Gloves
Description Packaged Individually
Disposable Single-use
Gender Unisex
Latex Free Yes
Neck Style Hooded
No. of Pockets None
Product Type Hooded Coverall
Quantity 6 Pk.
Resistance Liquid
Seam Type Taped
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.