Learn More
DuPont™ Tychem™ 6000 Coverall with Respirator Fit Hood
Catalog No. 18888796 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-888-796 4X-Large
18-888-797 5X-Large
18-888-799 Medium
18-888-800 Small
18-888-794 2X-Large
18-888-795 3X-Large
18-888-798 Large
18-888-801 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 18-888-796 Supplier DuPont™ Supplier No. TF199TGY4X0006WG
Add to Cart
Add to Cart
This item is not returnable. View return policy
6000 Coverall with respirator fit hood provide protection from chemical warfare agents and toxic industrial chemicals.
  • Certified to NFPA 1990 (NFPA 1992), 2022 edition
  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Hooded coverall with longer zipper that extends to chin for complete coverage of neck area and good fit around a respirator
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Attached Dual Layer Gloves, Internal: Multi-layer laminate / External: Butyl
  • Attached socks
  • Outer boot flaps
  • Manufactured under specifications that do not contain natural rubber latex
TRUSTED_SUSTAINABILITY
Specifications
Color Gray
Material Tychem
Size 4X-Large
Ankle Style Boot Overflaps
Closure Type Zipper
Cuff Style Attached Gloves
Disposable Single-use
Gender Unisex
Latex Free Yes
Neck Style Hooded
No. of Pockets None
Quantity 6 Pk.
Resistance Liquid
Seam Type Taped
Sterility Non-sterile