Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ 6000F Hooded Coverall with Rubber Face Seal

Catalog No. 18888808 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-888-808 Small
18-888-804 4X-Large
18-888-805 5X-Large
18-888-807 Medium
18-888-802 2X-Large
18-888-803 3X-Large
18-888-806 Large
18-888-809 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 18-888-808 Supplier DuPont™ Supplier No. TF611TGYSM000109
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Innovative features including a rubber tight seal around the mask and attached gloves for continued chemical protection from sleeve to glove

  • Tychem™ 6000 F FaceSeal, provides tighter chemical protection, enables longer wear time, and better comfort.
    • This unique fabric offers protection against both organic and highly concentrated inorganic chemicals
  • Lightweight, <500g per coverall
  • Rubber face seal for compatibility with full face masks, no taping needed
  • Rear entry with double flaps, press stud buttons and zipper for enhanced user protection
  • Attached dissipative socks with boot flap
  • High level chemical protection inner glove attached

Certifications

Cat. III, Type 3-B,4-B,5-B,6-B

Compliance

Meets ASTM™ D3776, ASTM D3787, ASTM D5733, ASTM D751, ASTM D2582; and 16 CFR 1610 Standard for Flammability of Clothing Textiles.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ASTM D3776, ASTM D3787, ASTM D5733, ASTM D751, ASTM D2582, 16 CFR 1610
Color Gray
For Use With (Application) Law Enforcement, Emergency Response, Industrial Cleaning
Hood Type Attached
Material Tychem
Size Small
Ankle Style Boot Overflaps
Cuff Style Attached Gloves
Disposable Single-use
Gender Unisex
Latex Free Yes
Neck Style Hooded
No. of Pockets None
Product Type Hooded Coverall
Quantity 1 Ea.
Resistance Liquid
Seam Type Taped
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.