Learn More
DuPont™ Tychem™ 6000F Hooded Coverall With Rubber Face Seal
Catalog No. p-7139632 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-888-804 4X-Large
18-888-805 5X-Large
18-888-807 Medium
18-888-808 Small
18-888-802 2X-Large
18-888-803 3X-Large
18-888-806 Large
18-888-809 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 18-888-804 Supplier DuPont™ Supplier No. TF611TGY4X000111
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Tychem™ 6000 F Hooded Coverall features several innovative features including a rubber tight seal around the mask and attached gloves for continued chemical protection from sleeve to glove.
Tychem™ 6000 F FaceSeal, provides tighter chemical protection, enables longer wear time, and better comfort. This unique fabric offers protection against both organic and highly concentrated inorganic chemicals.
  • Lightweight, <500g per coverall
  • Rubber face seal for compatibility with full face masks, no taping needed
  • Rear entry with double flaps, press stud buttons and zipper for enhanced user protection
  • Attached dissipative socks with boot flap
  • High level chemical protection inner glove attached
Certifications
Cat. III, Type 3-B,4-B,5-B,6-B
Compliance
Meets ASTM™ D3776, ASTM D3787, ASTM D5733, ASTM D751, ASTM D2582; and 16 CFR 1610 Standard for Flammability of Clothing Textiles.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ASTM D3776, ASTM D3787, ASTM D5733, ASTM D751, ASTM D2582, 16 CFR 1610
Color Gray
Material Tychem
Size 4X-Large
Ankle Style Boot Overflaps
Cuff Style Attached Gloves
Disposable Single-use
Gender Unisex
Latex Free Yes
Neck Style Hooded
No. of Pockets None
Quantity 1/Ea.
Resistance Liquid
Seam Type Taped
Sterility Non-sterile