Learn More
DuPont™ Tychem™ 6000 SFR FS275T LT Apron
Catalog No. 17500318 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-500-318 5X-Large
17-500-311 Small
17-500-312 Medium
17-500-313 Large
17-500-314 X-Large
17-500-315 2X-Large
17-500-316 3X-Large
17-500-317 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-500-318 Supplier DuPont™ Supplier No. FS275TLT5X001200
Add to Cart
Add to Cart
Tychem™ 6000 SFR Sleeved Apron. 44 in. Long. Hook and Loop Neck Closure. Waist Ties. Taped Seams. Light Tan.
  • Tychem™ 6000 SFR garments provide broad chemical protection and secondary flame resistance in a durable, lightweight garment for use in environments where chemical splash and flash fire hazards exist.
  • Designed to be worn over primary flame-resistant protective clothing, Tychem™ 6000 SFR garments help provide industrial workers and hazmat responders with permeation protection against a range of chemicals while providing escape time from flash fires.
  • Tychem™ 6000 SFR fabric provides at least 30 min. of protection for more than 250 chemical challenges including chemical warfare agents, toxic industrial chemicals, and flammable organic solvents.
  • In the event that a flash fire occurs, Tychem™ 6000 SFR garments won't contribute to additional burn injury if appropriate primary flame-resistant personal protective equipment is worn beneath.
TRUSTED_SUSTAINABILITY
Specifications
Type Apron
Size 5X-Large
Material Tychem
Color Tan
Disposable Single-use
Closure Type Hook and Loop, Waist Ties
Sterility Non-sterile
Latex Free No
No. of Pockets None
Seam Type Taped