Learn More
DuPont™ Tychem™ 6000 SFR FS750T LT Overalls
Catalog No. 17500319 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-500-319 Small
17-500-320 Medium
17-500-321 Large
17-500-322 X-Large
17-500-323 2X-Large
17-500-324 3X-Large
17-500-325 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 17-500-319 Supplier DuPont™ Supplier No. FS750TLTSM000400
Add to Cart
Add to Cart
Tychem™ 6000 SFR Bib Overall and Jacket Combo. Elastic Wrists. Open Ankles. Jacket has Double Storm Flap. Overalls have Adjustable Webbing Straps with Buckle Closure. Taped Seams. Light Tan.
  • Tychem™ 6000 SFR garments provide broad chemical protection and secondary flame resistance in a durable, lightweight garment for use in environments where chemical splash and flash fire hazards exist.
  • Designed to be worn over primary flame-resistant protective clothing, Tychem™ 6000 SFR garments help provide industrial workers and hazmat responders with permeation protection against a range of chemicals while providing escape time from flash fires.
  • Tychem™ 6000 SFR fabric provides at least 30 min. of protection for more than 250 chemical challenges including chemical warfare agents, toxic industrial chemicals, and flammable organic solvents.
  • In the event that a flash fire occurs, Tychem™ 6000 SFR garments won't contribute to additional burn injury if appropriate primary flame-resistant personal protective equipment, such as DuPont™ Nomex™, is worn beneath.
TRUSTED_SUSTAINABILITY
Specifications
Closure Type Buckle
Color Tan
Material Tychem
Seam Type Taped
Size Small
Suspender Adjustable
Ankle Style Open
No. of Pockets None
Product Type Overalls
Disposable Single-use