Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ 6000 SFR Light Tan Coverall FS127

Catalog No. 17500120 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-500-120 2X-Large
17-604-234 3X-Large
17-701-039 4X-Large
17-800-667 5X-Large
17-300-119 Large
17-200-856 Medium
17-111-300 Small
17-400-250 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-500-120 Supplier DuPont™ Supplier No. FS127TLT2X000600
Add to Cart
Add to Cart

Tychem™ 6000 SFR Coverall. Respirator Fit Hood. Front Zipper Closure. Chin Flap. Double Storm Flap. Elastic Face Opening. Elastic Wrists. Elastic Ankles. Elastic Waist. Taped Seams. Pin Lock Zipper. Light Tan.

  • Provide broad chemical protection and secondary flame resistance in a durable, lightweight garment for use in environments where chemical splash and flash fire hazards exist.
  • Designed to be worn over primary flame-resistant protective clothing, Tychem™ 6000 SFR garments help provide industrial workers and hazmat responders with permeation protection against a range of chemicals while providing escape time from flash fires.
  • Tychem™ 6000 SFR fabric provides at least 30 minutes of protection for more than 250 chemical challenges including chemical warfare agents, toxic industrial chemicals, and flammable organic solvents.
  • Garments won't contribute to additional burn injury if appropriate primary flame-resistant personal protective equipment, such as DuPont™ Nomex™, is worn beneath.
  • Applications include: oil and gas, industrial chemical processing plants, laboratories, hazmat teams in fire departments, industrial fire brigades, and clandestine lab investigation.
  • Respirator-fit hood
  • Covered elastic face opening
  • Covered elastic openings at wrists and ankles for tighter fit
  • Elastic at sides of waist for better fit
  • Chin flap with double-sided adhesive tape
  • Double storm flap covers zipper which can be sealed by the wearer with double-sided adhesive strip to prevent intrusion at zipper.
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Nylon zipper with large metal pull
  • Pin lock zipper prevents movement of the slider when puller is in the down position, helping to prevent garment from unzipping while performing work.
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks
TRUSTED_SUSTAINABILITY

Specifications

Product Type Protective Coveralls
Size 2X-Large
Gender Unisex
Material Tychem
Color Tan
Disposable Reusable
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Respirator Fit
Closure Type Zipper
Seam Type Taped
Sterility Non-sterile
Resistance Chemicals, Flame
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.