Description
DuPont™ Tychem™ 9000 is strong and durable for consistent and lasting protection.
- NFPA 1994 Class 2 and NFPA 1992 certified
- Provides at least 30 minutes of protection against over 280 chemical challenges
- Used by many in the chemical handling and petrochemical manufacturing
Specifications
Specifications
| Color | Yellow |
| Material | Santoprene |
| Size | 2X-Large |
| Ankle Style | Open ankles |
| Closure Type | Buckle closure |
| Seam Type | Taped |
| Style | Double storm flap jacket |
| Suspender | Adjustable webbing straps with buckle closure |