Learn More
DuPont™ Tychem™ 9000 Encapsulated Level A Suit
Catalog No. 17500026 Shop All DuPont Personal Protection Products
This item is not returnable. View return policy
This item is not returnable. View return policy
DuPont™ Tychem™ 9000 Encapsulated Level A Suit offers protection against a wide range of chemical challenges, resistance to tears, punctures, and abrasion.
DuPont™ Tychem™ 9000 is strong and durable for consistent and lasting protection.
  • NFPA 1994 Class 2 and NFPA 1992 certified
  • Provides at least 30 minutes of protection against over 280 chemical challenges
  • Used by many in the chemical handling and petrochemical manufacturing
  • Extra wide visor
  • 2 exhaust valves
  • Option 7R: MSA dual purpose passthrough with Hansen fitting
TRUSTED_SUSTAINABILITY
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.