Learn More
DuPont™ Tychem™ 9000 Encapsulated Level A Suit
Catalog No. p-7138983 Shop All DuPont Personal Protection Products
This item is not returnable. View return policy
Catalog No. 17-500-023 Supplier DuPont™ Supplier No. BR557TYL2X00017R
This item is not returnable. View return policy
DuPont™ Tychem™ 9000 Encapsulated Level A Suit offers protection against a wide range of chemical challenges, resistance to tears, punctures, and abrasion.
DuPont™ Tychem™ 9000 is strong and durable for consistent and lasting protection.
  • NFPA 1994 Class 2 and NFPA 1992 certified
  • Provides at least 30 minutes of protection against over 280 chemical challenges
  • Used by many in the chemical handling and petrochemical manufacturing
  • Extra wide visor
  • 2 exhaust valves
  • Option 7R: MSA dual purpose passthrough with Hansen fitting