Learn More
DuPont™ Tychem™ 9000 Encapsulated Level B Suit
Catalog No. 17500022 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Tychem™ 9000 Encapsulated Level B Suit
Catalog No.
17-500-022 Tychem™ 9000 Encapsulated Level B Suit
1 options
This item is not returnable. View return policy
Catalog No. 17-500-022 Supplier DuPont™ Supplier No. BR528TYL4X000100
This item is not returnable. View return policy
DuPont™ Tychem™ 9000 Encapsulated Level B Suit offers protection against a wide range of chemical challenges, resistance to tears, punctures, and abrasion.
DuPont™ Tychem™ 9000 is strong and durable for consistent and lasting protection.
  • NFPA 1994 Class 2 and NFPA 1992 certified
  • Provides at least 30 minutes of protection against over 280 chemical challenges
  • Used by many in the chemical handling and petrochemical manufacturing