Learn More
DuPont™ Tychem™ 5000 Encapsulated Level B
Catalog No. 190881810 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Ankle Style:
Attached Socks
Attached Socks And Boot Flaps
Size:
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Ankle Style Size
19-088-1810 Attached Socks And Boot Flaps 3X-Large
19-822-333 Attached Socks And Boot Flaps Large
19-059-236 Attached Socks 3X-Large
19-715-279 Attached Socks X-Large
19-088-1805 Attached Socks And Boot Flaps 2X-Large
19-059-238 Attached Socks 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-088-1810 Supplier DuPont™ Supplier No. C3528TTN3X000600
Add to Cart
Add to Cart
This item is not returnable. View return policy
Zipper and double storm flap with hook-and-loop closure
  • Encapsulated design provides protection from liquid splash exposure for both the wearer and respiratory equipment
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks
  • Also provides coverage from overhead liquid splash (does not provide vapor protection)
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
  • Standard 20 mil PVC faceshield
  • Rear entry design keeps closure away from direct contact with frontal chemical exposure
  • Flat back design for use with airline
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
TRUSTED_SUSTAINABILITY
Specifications
Color Tan
Material Tychem
Size 3X-Large
Ankle Style Attached Socks And Boot Flaps
Closure Type Zipper
Seam Type Taped