Learn More
Description
- Encapsulated design provides protection from liquid splash exposure for both the wearer and respiratory equipment
- Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
- Manufactured under specifications that do not contain natural rubber latex
- Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks
- Provides coverage from overhead liquid splash (does not provide vapor protection)
- Taped seams provide strong chemical resistance against heavy liquid splashes
- Sewn seam covered with a strip of compatible chemical-resistant material through heat sealing
- Standard 20 mil PVC face shield
- Rear entry design keeps closure away from direct contact with frontal chemical exposure
- Flat back design for use with airline
- Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
- Elastic opening for tighter fit at wrist
- Integrated socks composed of garment material
Specifications
Specifications
| Color | Tan |
| For Use With (Application) | Chemical Handling, Hazmat, Law Enforcement, Domestic Preparedness |
| Material | Tychem |
| Size | Large |
| Ankle Style | Attached Socks and Boot Flaps |
| Closure Type | Zipper |
| Product Type | Encapsulated Level B Suit |
| Seam Type | Taped |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.