Learn More
DuPont™ Tychem™ PV350 Black PVC Gloves
Catalog No. 19715407 Shop All DuPont Personal Protection Products
Change view
Click to view available options
:
Tychem™ PV350 Glove
Catalog No.
19-715-407 Tychem™ PV350 Glove
1 options
This item is not returnable. View return policy
Catalog No. 19-715-407 Supplier DuPont™ Supplier No. PV3500BK100144CF
This item is not returnable. View return policy
Black, cotton-jersey lined PVC glove provides defensive protection from chemicals, oil and grease.
  • PVC with cotton/jersey lining
  • Suitable for general-purpose industrial applications
  • Durable, thick
  • Barrier protection against chemicals, oils and greases
  • Abrasion resistance
  • Versatile
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) Cat III CE 2777
Chlorinated No
Color Black
Hand Specific
Latex Free Yes
Material PVC
Size 10
Thickness (English) 55 mil
Thickness (Metric) 1.40 mm
EN388 Standard EN 388:3111X
EN 421 No
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES AKLMP
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Gauntlet
Grip Design Rough
Liner Material Cotton/Jersey
Lining Lined
Model PV3500BK100144CF
Series Tychem™ PV350
Length (English) 14 in.
Length (Metric) 35.6 cm