Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ Tychem QC Double Bib Neck Cape Assembly
SDP

Catalog No. 18444000
Change view
Click to view available options
Model:
433551
433551 SP
Quantity:
1 Ea.
40/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Quantity
18-444-000 433551 1 Ea.
18-444-001 433551 SP 40/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-444-000 Supplier Gentex™ Supplier No. 433551
Add to Cart
Add to Cart
This item is not returnable. View return policy

Tychem QC double bib neck cape assembly for use with PureFlo ESM and ESM+ PAPR systems.

  • Tychem 2000 (QC) fabric consists of Tyvek brand fabric with polyethylene coating
  • Tested by third party laboratory to assess chemical penetration resistance
  • Protects against light liquid splash
  • Used for the following applications:
    • Petroleum
    • Paper and pulp
    • Food and chemical processing
    • Pharmaceutical
    • Biological
  • Used with PureFlo ESM and ESM+ PAPR systems.

Specifications

Color White
Disposable Single-use
Material Tychem
Product Type Neck Cape
Size One Size
For Use With (Equipment) PureFlo ESM and ESM+ PAPR Systems
Latex Free Yes
Model 433551
Quantity 1 Ea.
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.