Learn More
DuPont™ Tychem™ RESPONDER™ CSM Encapsulated Level A Suit
Catalog No. 18888739 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
X-Small
Small
5X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-888-739 5X-Large
18-888-747 Small
18-888-753 X-Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 18-888-739 Supplier DuPont™ Supplier No. RC550TTN5X000100
Add to Cart
Add to Cart
This item is not returnable. View return policy
CSM Encapsulated Suit is a low visibility, front entry suit with attached gloves and socks
  • CSM Encapsulated Level A Suit design is our highest level of protection from liquid splash and vapor/gas exposures for both the wearer and respiratory equipment
  • Each suit has a unique serial number and is fully tested at time of manufacture including positive air pressure integrity testing per ASTM F1052.
  • Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
  • Standard-width, anti-fog, three-layer (PVC 40 mil / Teflon™ 5 mil / PVC 20 mil) faceshield
  • Front entry design allows wearer to partially don garment (maintain standby position) without assistance
  • Expanded back to accommodate self-contained air breathing apparatus (SCBA)
  • Vapor tight zipper
  • Extra long to increase garment opening and aid donning and doffing
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • Integrated socks composed of garment material
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
TRUSTED_SUSTAINABILITY
Specifications
Color Tan
Material Tychem
Size 5X-Large
Ankle Style Boot Overflaps
Closure Type Zipper
Seam Type Taped