Learn More
DuPont™ Tychem™ RESPONDER™ CSM Encapsulated Level A Suit with Hansen Fitting
Shop All DuPont Personal Protection ProductsDescription
- CSM Encapsulated Level A Suit with Hansen Fitting design is our highest level of protection from liquid splash and vapor/gas exposures for both the wearer and respiratory equipment
- Each suit has a unique serial number and is fully tested at time of manufacture including positive air pressure integrity testing per ASTM F1052
- Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
- Double taped seams provide strong chemical resistance against heavy liquid splashes
- Sewn seam is covered, both inside and outside of garment with compatible chemical-resistant material by heat-sealing
- Standard-width, anti-fog, three-layer (PVC 40 mil, Teflon™ 5 mil, PVC 20 mil) face shield
- Front entry design allows wearer to partially don garment (maintain standby position) without assistance
- Expanded back to accommodate self-contained air breathing apparatus (SCBA)
- Vapor tight zipper. Extra long to increase garment opening and aid donning and doffing
- Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
- Integrated socks composed of garment material
- Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
Specifications
Specifications
| Color | Tan |
| For Use With (Application) | Military Site Cleanup, Hazmat First Response |
| Material | Tychem |
| Size | X-Large |
| Ankle Style | Boot Overflaps |
| Closure Type | Zipper |
| Product Type | Encapsulated Level A Suit |
| Seam Type | Taped |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.