Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ RESPONDER™ CSM Encapsulated Level A Suit with MSA CAMDS Connector

Catalog No. 18888750 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-888-750 X-Large
18-888-730 2X-Large
18-888-733 3X-Large
18-888-736 4X-Large
18-888-740 5X-Large
18-888-741 Large
18-888-744 Medium
18-888-748 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 18-888-750 Supplier DuPont™ Supplier No. RC550TTNXL00017C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Front entry, low visibility suit with attached socks and gloves

  • CSM Encapsulated Level A Suit with MSA CAMDS Connector design is our highest level of protection from liquid splash and vapor/gas exposures for both the wearer and respiratory equipment
  • Two exhaust valves release higher pressure inside encapsulating garments while simultaneously helping to prevent inward leakage of external vapors or particles into the garment
    • One-way valves are positive pressure and open on demand
    • Include splash covers made of barrier material to help prevent liquid intrusion
    • Located on left back of head and right lower back
  • Each suit has a unique serial number and is fully tested at time of manufacture including positive air pressure integrity testing per ASTM F1052
  • Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
  • Double taped seams provide strong chemical resistance against heavy liquid splashes
    • Sewn seam is covered, both inside and outside of garment with compatible chemical-resistant material by heat-sealing.
  • Standard-width, anti-fog, three-layer (PVC 40 mil, Teflon™ 5 mil, PVC 20 mil) face shield
  • Front entry design allows wearer to partially don garment (maintain standby position) without assistance
  • Expanded back to accommodate self-contained air breathing apparatus (SCBA)
  • Vapor tight zipper
    • Extra long to increase garment opening and aid donning and doffing
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • Integrated socks composed of garment material
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
TRUSTED_SUSTAINABILITY

Specifications

Color Tan
For Use With (Application) Military Site Cleanup, Hazmat First Response
Material Tychem
Size X-Large
Ankle Style Boot Overflaps
Closure Type Zipper
Product Type Encapsulated Level A Suit
Seam Type Taped
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.