Learn More
DuPont™ Tychem™ ThermoPro™ Sleeveless Gray Aprons
Catalog No. 19122003 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-122-003 X-Large
19-122-000 Small
19-122-001 Medium
19-122-002 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-122-003 Supplier DuPont™ Supplier No. TP284TGYXL000400
This item is not returnable. View return policy
Designed to provide protection for industrial workers and Hazmat responders. DuPont™ Tychem™ ThermoPro™ Sleeveless Aprons provide permeation protection against a broad range of toxic industrial chemicals and chemical warfare agents
  • Tychem™ ThermoPro for triple hazard protection
  • Sleeveless apron design provides frontal body protection
  • For flash fire and electric arc hazards, partial body protective Tychem ThermoPro garments must be used in conjunction with primary flame resistant clothing that is rated for the fire/arc hazard.
  • Sewn with Nomex™ thread
  • Designed with Nomex waist strap and flame resistant (FR) nylon buckle
  • Designed with end-user input for laboratory applications
  • Ideal for use on top of an existing flame resistant (FR) garment where extra chemical protection is required
  • Gray color for discretionary needs
  • Made in the USA
Order Info
Ansell™ Vinyl Closed-Back Aprons, Waist ties
Compliance
North American Free Trade Agreement (NAFTA) and Trade Agreement Act (TAA).
TRUSTED_SUSTAINABILITY
Specifications
Size X-Large
Color Gray
For Use With (Application) HazMat Teams in Fire Departments, Industrial Fire Brigades, Clandestine Lab Investigation, Industrial Chemical Processing Plants, Laboratories
Seam Type Taped