Learn More
MSC TYCHEM5000 BIB OVRL/JCK
Catalog No. 5000802716
Encompass
The DuPont Tychem 5000 Bib Overall & Jacket Combo provides chemical resistance and durability in hazardous environments. This ensemble offers a barrier against a broad range of chemicals and liquid splash for industrial and emergency response.
  • Fabric: multi-layer composite barrier film laminated to polypropylene.
  • Bib overall and jacket combination with mandarin collar, jam fit cuffs, and open ankles.
  • Jacket features a front zipper with double storm flap and hook and loop closure.
  • Taped seams provide chemical resistance.
  • Meets U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671).
  • Manufactured without natural rubber latex and silicone.
  • Ambient working temperature range: -13°F (-25°C) to 165°F (74°C).
  • Suitable for chemical handling, petrochemical, and hazardous waste cleanup.
Catalog No. 50-008-02716 Supplier MSC Supplier No. 65431074
The DuPont Tychem 5000 Bib Overall & Jacket Combo provides chemical resistance and durability in hazardous environments. This ensemble offers a barrier against a broad range of chemicals and liquid splash for industrial and emergency response.
  • Fabric: multi-layer composite barrier film laminated to polypropylene.
  • Bib overall and jacket combination with mandarin collar, jam fit cuffs, and open ankles.
  • Jacket features a front zipper with double storm flap and hook and loop closure.
  • Taped seams provide chemical resistance.
  • Meets U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671).
  • Manufactured without natural rubber latex and silicone.
  • Ambient working temperature range: -13°F (-25°C) to 165°F (74°C).
  • Suitable for chemical handling, petrochemical, and hazardous waste cleanup.