Learn More
MSC TYCHEM5000 COLLAR JACKET
Catalog No. 5000803215
Encompass
The Tychem 5000 Jacket provides heavy-duty protection against a wide variety of chemical hazards. This tan jacket extends to the hip and features a mandarin collar, elastic wrists, a double storm flap with hook and loop closure, and taped seams. It is suitable for chemical handling, hazmat, law enforcement, and domestic preparedness applications.
  • Fabric is a multi-layer film barrier laminated to a durable 2.0 oz/yd² polypropylene substrate.
  • Strong chemical resistance against heavy liquid splashes due to taped seams.
  • Mandarin collar fits tightly around hooded PAPR.
  • Storm flap covers zipper with hook and loop closure.
  • Manufactured without natural rubber latex.
  • Meets U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671).
Catalog No. 50-008-03215 Supplier MSC Supplier No. 53841128
The Tychem 5000 Jacket provides heavy-duty protection against a wide variety of chemical hazards. This tan jacket extends to the hip and features a mandarin collar, elastic wrists, a double storm flap with hook and loop closure, and taped seams. It is suitable for chemical handling, hazmat, law enforcement, and domestic preparedness applications.
  • Fabric is a multi-layer film barrier laminated to a durable 2.0 oz/yd² polypropylene substrate.
  • Strong chemical resistance against heavy liquid splashes due to taped seams.
  • Mandarin collar fits tightly around hooded PAPR.
  • Storm flap covers zipper with hook and loop closure.
  • Manufactured without natural rubber latex.
  • Meets U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671).