Learn More
MSC Dupont Hooded Coverall, Tychem 5000, X-Large, Tan, 6/Case
Catalog No. 5000789371
Encompass
The DuPont™ Tychem® 5000 Hooded Coverall offers heavy-duty protection against various chemical hazards. Featuring a standard hood, elastic wrists and ankles, and taped seams, it is ideal for chemical handling, hazmat, and domestic preparedness.
  • Multi-layer film barrier fabric (2.0 oz/yd² polypropylene) provides robust chemical resistance.
  • Strong, durable design for rigorous activities involving liquid splash.
  • Semi-automatic locking slider zipper with storm flap and adhesive closure.
  • Taped seams ensure strong chemical resistance against heavy liquid splashes.
  • Elastic wrists, ankles, and hood opening for a secure fit.
  • Meets U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671).
Catalog No. 50-007-89371 Supplier MSC Supplier No. 09543265
The DuPont™ Tychem® 5000 Hooded Coverall offers heavy-duty protection against various chemical hazards. Featuring a standard hood, elastic wrists and ankles, and taped seams, it is ideal for chemical handling, hazmat, and domestic preparedness.
  • Multi-layer film barrier fabric (2.0 oz/yd² polypropylene) provides robust chemical resistance.
  • Strong, durable design for rigorous activities involving liquid splash.
  • Semi-automatic locking slider zipper with storm flap and adhesive closure.
  • Taped seams ensure strong chemical resistance against heavy liquid splashes.
  • Elastic wrists, ankles, and hood opening for a secure fit.
  • Meets U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671).