Learn More
US Plastic Corporation 5/16" ID x 9/16" OD x 1/8" Wall Tygon S3 E-3603 Laboratory Tubing, 50ft
SDP
Catalog No. NC2006236
Encompass
This tubing is ideal for general laboratory use, analytical instruments, peristaltic pumps, condensers, incubators, desiccators, as well as gas and drain lines. This tubing offers superior life and minimizes labor and the expense of replacement. It is non-oxidizing and non-contaminating, and less permeable than rubber tubing.
Catalog No. NC2006236 Supplier US Plastic Corporation Supplier No. 5764250FT
This tubing is ideal for general laboratory use, analytical instruments, peristaltic pumps, condensers, incubators, desiccators, as well as gas and drain lines. This tubing offers superior life and minimizes labor and the expense of replacement. It is non-oxidizing and non-contaminating, and less permeable than rubber tubing.