Learn More
Accuflow Systems Saint-Gobain Tygon S3 Tubing, TPE, 1/2 in ID, 50 ft
Catalog No. NC3713029
Encompass_Preferred
Saint-Gobain Tygon S3 B-44-5X IB is a reinforced thermoplastic elastomer dispensing tubing formulated for food and beverage applications requiring elevated pressure and hygienic bore characteristics. The tubing features a smooth, non-porous inner bore to inhibit particle entrapment, is virtually taste- and odor-free, and resists common sanitizers and alkaline cleaners.
  • Reinforced construction for elevated working pressure and routing flexibility.
  • Smooth, non-porous inner bore to reduce particle entrapment and bacterial growth.
  • TPE material with resistance to common sanitizers and alkaline cleaning solutions.
  • Wide temperature range for frozen beverage and dispensing equipment use.
  • Complete drainage and non-wetting inner surface for easy flush-cleaning.
Catalog No. NC3713029 Supplier Accuflow Systems Supplier No. AMH00038
Saint-Gobain Tygon S3 B-44-5X IB is a reinforced thermoplastic elastomer dispensing tubing formulated for food and beverage applications requiring elevated pressure and hygienic bore characteristics. The tubing features a smooth, non-porous inner bore to inhibit particle entrapment, is virtually taste- and odor-free, and resists common sanitizers and alkaline cleaners.
  • Reinforced construction for elevated working pressure and routing flexibility.
  • Smooth, non-porous inner bore to reduce particle entrapment and bacterial growth.
  • TPE material with resistance to common sanitizers and alkaline cleaning solutions.
  • Wide temperature range for frozen beverage and dispensing equipment use.
  • Complete drainage and non-wetting inner surface for easy flush-cleaning.