Learn More
Accuflow Systems Saint-Gobain Tygon Microbore Tubing, Non-DEHP, 0.010 in ID, 500 ft
Catalog No. 502365830
Encompass_Preferred
Tygon ND 100-80 microbore tubing is a non-DEHP medical-grade thermoplastic designed for precise microfluidic and medical device fluid transfer. It offers Class VI biocompatibility and a higher durometer for improved pressure resistance at very small diameters.
  • Non-DEHP medical thermoplastic
  • 0.010 in ID x 0.030 in OD, 0.010 in wall (0.25 mm x 0.75 mm, 0.25 mm)
  • Packaged length 500 ft (150 m)
  • Meets USP <88> Biological Test for Plastics, Class VI (fluid path)
  • Suitable for drainage systems, diagnostic equipment, and pressurized fluid transfer
  • Microbore size compatible with peristaltic pumps and analytical instrumentation
Catalog No. 50-236-5830 Supplier Accuflow Systems Supplier No. AAD04091
Tygon ND 100-80 microbore tubing is a non-DEHP medical-grade thermoplastic designed for precise microfluidic and medical device fluid transfer. It offers Class VI biocompatibility and a higher durometer for improved pressure resistance at very small diameters.
  • Non-DEHP medical thermoplastic
  • 0.010 in ID x 0.030 in OD, 0.010 in wall (0.25 mm x 0.75 mm, 0.25 mm)
  • Packaged length 500 ft (150 m)
  • Meets USP <88> Biological Test for Plastics, Class VI (fluid path)
  • Suitable for drainage systems, diagnostic equipment, and pressurized fluid transfer
  • Microbore size compatible with peristaltic pumps and analytical instrumentation