Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuflow Systems Saint-Gobain Tygon Tubing, Non-DEHP, 0.02 in ID, 500 ft

Catalog No. 502365803
Encompass_Preferred

Tygon ND 100-80 is a non-DEHP, medical-grade microbore thermoplastic tubing designed for precise fluid transfer in medical, diagnostic, and drug delivery applications. It offers a relatively high durometer for increased pressure resistance while permitting small diameters and thin walls, and it tolerates common sterilization methods without significant changes to physical properties.

  • Inner diameter 0.02 in (0.51 mm).
  • Outer diameter 0.06 in (1.52 mm).
  • Wall thickness 0.02 in (0.505 mm).
  • Package length 500 ft (152 m).
  • Fluid path meets USP <88> (2017) Biological Test for Plastics, Class VI.
  • Compatible with autoclave (steam at 135°C for 30 min), ethylene oxide, and gamma sterilization without significant change to physical properties.
  • Suitable for intravenous infusion, cannulas, external drainage, diagnostic reagent transfer, and pressurized fluid transfer.

Catalog No. 50-236-5803 Supplier Accuflow Systems Supplier No. AAD04103
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tygon ND 100-80 is a non-DEHP, medical-grade microbore thermoplastic tubing designed for precise fluid transfer in medical, diagnostic, and drug delivery applications. It offers a relatively high durometer for increased pressure resistance while permitting small diameters and thin walls, and it tolerates common sterilization methods without significant changes to physical properties.

  • Inner diameter 0.02 in (0.51 mm).
  • Outer diameter 0.06 in (1.52 mm).
  • Wall thickness 0.02 in (0.505 mm).
  • Package length 500 ft (152 m).
  • Fluid path meets USP <88> (2017) Biological Test for Plastics, Class VI.
  • Compatible with autoclave (steam at 135°C for 30 min), ethylene oxide, and gamma sterilization without significant change to physical properties.
  • Suitable for intravenous infusion, cannulas, external drainage, diagnostic reagent transfer, and pressurized fluid transfer.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.