Learn More
Saint Gobain Performance Plastics Tygon S3™ Tubing | 1/32" I.D. x 3/32" O.D. x 1/32" Wall | 50' Length per package.
Supplier:  Saint Gobain Performance Plastics ACF00001
Encompass_Preferred
This item has a minimum order quantity of 50 packs (2500ft) per supplier requirementsSaint-Gobain is proud to be among the first companies to offer sustainable flexible tubing products. The bio-based Tygon S3 line combines the high performance standards customers demand with an eco-friendly tubing design. Tygon S3 E-3603 is crystal clear and flexible, long-lasting and crack-resistant. Temp Range -51 deg. F (-46 deg. C) - 165 deg. F (74 deg. C)
Catalog No. 50-206-8934