Learn More
Saint Gobain Performance Plastics Tube Tygon 1/4 x 7/16 50FT-15M/PK
Supplier:  Saint Gobain Performance Plastics ACF00018
Encompass_Preferred
Saint-GobainTygon S3 E-3603 Flexible Tubings Diameter Inner 1/4 in. Diameter Outer 7/16 in. Thickness 3/32 in. Overall Length 50 ft. 15 m Clear
Catalog No. 50-206-8947