Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, 1/16 in I.D./1/8 in O.D., 50 ft
Catalog No. 502365939
Encompass_Preferred
Tygon E-3603 tubing is a crystal-clear, flexible thermoplastic tubing formulated for medical and diagnostic fluid handling where visual inspection, chemical compatibility, and long service life are required. The tubing's high dimensional tolerance and resistance to flex-fatigue help provide consistent, reproducible fluid transfer and minimize downtime in analytical and instrumentation applications.
  • Flexible, crystal-clear thermoplastic for visual inspection and fluid transfer.
  • 1/16 in inner diameter, 1/8 in outer diameter, 1/32 in wall, 50 ft length.
  • Glassy-smooth inner bore resists buildup and reduces carryover.
  • Non-oxidizing, non-contaminating formulation minimizes extractables.
  • High resistance to flex-fatigue and abrasion for extended service life.
  • Suitable for laboratory, medical diagnostic, and instrumentation applications.
Catalog No. 50-236-5939 Supplier Accuflow Systems Supplier No. ACF00002F
Tygon E-3603 tubing is a crystal-clear, flexible thermoplastic tubing formulated for medical and diagnostic fluid handling where visual inspection, chemical compatibility, and long service life are required. The tubing's high dimensional tolerance and resistance to flex-fatigue help provide consistent, reproducible fluid transfer and minimize downtime in analytical and instrumentation applications.
  • Flexible, crystal-clear thermoplastic for visual inspection and fluid transfer.
  • 1/16 in inner diameter, 1/8 in outer diameter, 1/32 in wall, 50 ft length.
  • Glassy-smooth inner bore resists buildup and reduces carryover.
  • Non-oxidizing, non-contaminating formulation minimizes extractables.
  • High resistance to flex-fatigue and abrasion for extended service life.
  • Suitable for laboratory, medical diagnostic, and instrumentation applications.