Learn More
Accuflow Systems Saint-Gobain Tygon S3 Tubing, PVC, 1/2 in ID, 50 ft
Catalog No. 502365790
Encompass_Preferred
Saint-Gobain Tygon S3 tubing is a clear, flexible PVC tubing formulated with a bio-based, non-DEHP plasticizer. It features extra-heavy walls for vacuum resistance and long-term durability, and is supplied in 50 ft (15 m) lengths with a 1/2 in (12.7 mm) inner diameter and 3/32 in (2.4 mm) wall thickness.
  • Clear PVC tubing with bio-based, non-DEHP plasticizer.
  • 1/2 in (12.7 mm) inner diameter, 11/16 in (17.5 mm) outer diameter, 3/32 in (2.4 mm) wall thickness.
  • Supplied in 50 ft (15 m) coils for extended runs.
  • Extra-heavy wall construction resists collapse under vacuum.
  • Suitable for laboratory, analytical, biopharmaceutical, and food contact applications.
  • Complies with FDA food contact and NSF 51 per manufacturer documentation.
Catalog No. 50-236-5790 Supplier Accuflow Systems Supplier No. ACF00037
Saint-Gobain Tygon S3 tubing is a clear, flexible PVC tubing formulated with a bio-based, non-DEHP plasticizer. It features extra-heavy walls for vacuum resistance and long-term durability, and is supplied in 50 ft (15 m) lengths with a 1/2 in (12.7 mm) inner diameter and 3/32 in (2.4 mm) wall thickness.
  • Clear PVC tubing with bio-based, non-DEHP plasticizer.
  • 1/2 in (12.7 mm) inner diameter, 11/16 in (17.5 mm) outer diameter, 3/32 in (2.4 mm) wall thickness.
  • Supplied in 50 ft (15 m) coils for extended runs.
  • Extra-heavy wall construction resists collapse under vacuum.
  • Suitable for laboratory, analytical, biopharmaceutical, and food contact applications.
  • Complies with FDA food contact and NSF 51 per manufacturer documentation.