Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuflow Systems Saint-Gobain Tygon Tubing, 1/2 in ID, 3/4 OD, 50 ft

Catalog No. 502365885
Encompass_Preferred

Saint-Gobain Tygon E-3603 tubing is a crystal-clear, flexible thermoplastic designed for reliable fluid transfer in medical, diagnostic, and laboratory instrumentation. The material offers low extractables, high dimensional tolerance, and resistance to abrasion and flex-fatigue for reproducible performance and easy visual inspection.

  • Clear, glassy-smooth inner bore reduces buildup and carryover.
  • Flexible construction with resistance to flex-fatigue for tight bends.
  • Non-oxidizing, non-contaminating formulation preserves sensitive reagents.
  • Dimensions: 1/2 in (12.7 mm) ID, 3/4 in (19.1 mm) OD, 1/8 in (3.2 mm) wall, 50 ft (15 m) length.
  • Maximum recommended operating temperature 165°F (74°C).
  • Shore A hardness 56; tensile strength 1,750 psi (12.1 MPa); specific gravity 1.21.

Catalog No. 50-236-5885 Supplier Accuflow Systems Supplier No. ACF00038
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Saint-Gobain Tygon E-3603 tubing is a crystal-clear, flexible thermoplastic designed for reliable fluid transfer in medical, diagnostic, and laboratory instrumentation. The material offers low extractables, high dimensional tolerance, and resistance to abrasion and flex-fatigue for reproducible performance and easy visual inspection.

  • Clear, glassy-smooth inner bore reduces buildup and carryover.
  • Flexible construction with resistance to flex-fatigue for tight bends.
  • Non-oxidizing, non-contaminating formulation preserves sensitive reagents.
  • Dimensions: 1/2 in (12.7 mm) ID, 3/4 in (19.1 mm) OD, 1/8 in (3.2 mm) wall, 50 ft (15 m) length.
  • Maximum recommended operating temperature 165°F (74°C).
  • Shore A hardness 56; tensile strength 1,750 psi (12.1 MPa); specific gravity 1.21.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.