Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, 1/2-in ID, 5/8-in OD, 50 ft
Catalog No. 502365850
Encompass_Preferred
Saint-Gobain Tygon E-3603 tubing is a crystal-clear, phthalate- and BPA-free thermoplastic tubing formulated for laboratory and medical fluid handling applications where chemical compatibility, visual inspection, and low extractables are required.
  • Crystal-clear thermoplastic for visual inspection and bubble detection.
  • Phthalate- and BPA-free formulation reduces extractables and contamination risk.
  • 1/2 in (12.7 mm) inner diameter, 5/8 in (15.9 mm) outer diameter, 1/16 in (1.6 mm) wall.
  • Available in 50 ft (15 m) standard length for laboratory use.
  • Good chemical resistance and non-oxidizing for many inorganic reagents.
  • Maintains vacuum integrity at room temperature in standard laboratory sizes.
Catalog No. 50-236-5850 Supplier Accuflow Systems Supplier No. ACF00036
Saint-Gobain Tygon E-3603 tubing is a crystal-clear, phthalate- and BPA-free thermoplastic tubing formulated for laboratory and medical fluid handling applications where chemical compatibility, visual inspection, and low extractables are required.
  • Crystal-clear thermoplastic for visual inspection and bubble detection.
  • Phthalate- and BPA-free formulation reduces extractables and contamination risk.
  • 1/2 in (12.7 mm) inner diameter, 5/8 in (15.9 mm) outer diameter, 1/16 in (1.6 mm) wall.
  • Available in 50 ft (15 m) standard length for laboratory use.
  • Good chemical resistance and non-oxidizing for many inorganic reagents.
  • Maintains vacuum integrity at room temperature in standard laboratory sizes.