Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, PVC, 1/4 in ID, 50 ft
Catalog No. 502365863
Encompass_Preferred
Flexible, extra-heavy-wall thermoplastic tubing designed for laboratory and diagnostic fluid transfer and vacuum applications. The tubing provides vacuum integrity, clear visual inspection, and a non-contaminating, phthalate-free formulation suitable for sensitive applications.
  • Extra-heavy-wall thermoplastic tubing for vacuum integrity and flex resistance.
  • Clear, non-oxidizing, non-contaminating formulation for visual inspection.
  • 1/4 in inner diameter, 1/2 in outer diameter, 1/8 in wall thickness, 50 ft length.
  • Withstands full vacuum at room temperature (29.9 inHg / 760 mmHg).
  • Maximum recommended operating temperature 165 °F (74 °C); brittle temperature -51 °F (-46 °C).
  • Phthalate-free and BPA-free formulation to minimize contamination risk.
  • Suitable for laboratory, medical, diagnostic, and general fluid-handling applications.
Catalog No. 50-236-5863 Supplier Accuflow Systems Supplier No. ACF00019F
Flexible, extra-heavy-wall thermoplastic tubing designed for laboratory and diagnostic fluid transfer and vacuum applications. The tubing provides vacuum integrity, clear visual inspection, and a non-contaminating, phthalate-free formulation suitable for sensitive applications.
  • Extra-heavy-wall thermoplastic tubing for vacuum integrity and flex resistance.
  • Clear, non-oxidizing, non-contaminating formulation for visual inspection.
  • 1/4 in inner diameter, 1/2 in outer diameter, 1/8 in wall thickness, 50 ft length.
  • Withstands full vacuum at room temperature (29.9 inHg / 760 mmHg).
  • Maximum recommended operating temperature 165 °F (74 °C); brittle temperature -51 °F (-46 °C).
  • Phthalate-free and BPA-free formulation to minimize contamination risk.
  • Suitable for laboratory, medical, diagnostic, and general fluid-handling applications.