Learn More
Accuflow Systems Saint-Gobain Tygon E-3603 Tubing, 1/4 in ID, 7/16 in OD
Catalog No. 502365892
Encompass_Preferred
Saint-Gobain Tygon S3 E-3603 tubing is a flexible, high-clarity thermoplastic tubing formulated for laboratory, medical, and diagnostic fluid handling where chemical compatibility and visual inspection are required. The E-3603 formulation is phthalate-free (non-DEHP), non-oxidizing, and features a glassy-smooth inner bore to minimize buildup and carryover. The tubing is resistant to flex-fatigue and supports vacuum service in standard sizes, making it suitable for peristaltic pump and instrumentation applications.
  • Clear, high-clarity thermoplastic for visual inspection.
  • 1/4 in internal diameter, 7/16 in outer diameter, 3/32 in wall.
  • 50 ft standard package length.
  • Phthalate-free formulation to minimize extractables.
  • Glassy-smooth inner bore reduces buildup and carryover.
  • Resistant to flex-fatigue and compatible with vacuum service.
  • Suitable for medical, diagnostic, and laboratory fluid transfer.
Catalog No. 50-236-5892 Supplier Accuflow Systems Supplier No. ACF00018
Saint-Gobain Tygon S3 E-3603 tubing is a flexible, high-clarity thermoplastic tubing formulated for laboratory, medical, and diagnostic fluid handling where chemical compatibility and visual inspection are required. The E-3603 formulation is phthalate-free (non-DEHP), non-oxidizing, and features a glassy-smooth inner bore to minimize buildup and carryover. The tubing is resistant to flex-fatigue and supports vacuum service in standard sizes, making it suitable for peristaltic pump and instrumentation applications.
  • Clear, high-clarity thermoplastic for visual inspection.
  • 1/4 in internal diameter, 7/16 in outer diameter, 3/32 in wall.
  • 50 ft standard package length.
  • Phthalate-free formulation to minimize extractables.
  • Glassy-smooth inner bore reduces buildup and carryover.
  • Resistant to flex-fatigue and compatible with vacuum service.
  • Suitable for medical, diagnostic, and laboratory fluid transfer.