Saint-Gobain Tygon Power F-4040-A tubing is a flexible, fuel- and lubricant-resistant PVC tubing formulated to resist swelling and embrittlement from hydrocarbon-based fluids. Its translucent yellow construction allows visual monitoring of flow while providing UV and ozone resistance and easy routing for small engines, automotive, and equipment fluid-transfer applications.
- Fuel- and lubricant-resistant special PVC formulation
- 1/8 in inside diameter, 1/4 in outside diameter, 1/16 in wall
- 50 ft pre-cut length for inventory convenience
- Maximum working pressure 50 psi at 73°F
- Translucent yellow for fluid visibility and identification
- Resists swelling, hardening, and embrittlement from hydrocarbons
- Operating temperature up to 165°F (74°C), brittle to -35°F (-37°C)
- Highly flexible for tight-routing and easy installation