Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, Thermoplastic, 1 in ID, 1-1/4 in OD, 50 ft
Catalog No. 502365819
Encompass_Preferred
Tygon E-3603 thermoplastic tubing provides crystal-clear, medical-grade fluid transfer with a glassy-smooth inner bore to minimize buildup. The tubing is non-oxidizing, non-contaminating, and free of BPA and phthalates. This coil is supplied in 50 ft lengths and is suited for laboratory, diagnostic, and medical fluid-handling applications.
  • Clear thermoplastic tubing, 1 in inside diameter, 1-1/4 in outside diameter.
  • 1/8 in wall thickness, 50 ft coil length.
  • Glassy-smooth inner bore reduces buildup and facilitates visual inspection.
  • Non-oxidizing and non-contaminating, BPA- and phthalate-free.
  • Maximum working pressure 12 psi at 73°F (23°C); vacuum rating 5 inHg (127 mmHg).
  • Minimum bend radius 4-3/4 in; maximum recommended operating temperature 165°F (74°C).
Catalog No. 50-236-5819 Supplier Accuflow Systems Supplier No. ACF00062
Tygon E-3603 thermoplastic tubing provides crystal-clear, medical-grade fluid transfer with a glassy-smooth inner bore to minimize buildup. The tubing is non-oxidizing, non-contaminating, and free of BPA and phthalates. This coil is supplied in 50 ft lengths and is suited for laboratory, diagnostic, and medical fluid-handling applications.
  • Clear thermoplastic tubing, 1 in inside diameter, 1-1/4 in outside diameter.
  • 1/8 in wall thickness, 50 ft coil length.
  • Glassy-smooth inner bore reduces buildup and facilitates visual inspection.
  • Non-oxidizing and non-contaminating, BPA- and phthalate-free.
  • Maximum working pressure 12 psi at 73°F (23°C); vacuum rating 5 inHg (127 mmHg).
  • Minimum bend radius 4-3/4 in; maximum recommended operating temperature 165°F (74°C).