Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, Fuel Lubricant, 3/16 in ID, 50 ft
Catalog No. 502365778
Encompass_Preferred
Flexible fuel and lubricant tubing formulated to resist swelling and hardening from hydrocarbon-based fluids. Suitable for small-engine fuel lines and general fuel and lubricant transfer applications.
  • 3/16 in inner diameter, 5/16 in outer diameter.
  • 1/16 in wall thickness.
  • 50 ft length, bulk coil.
  • Translucent yellow for fluid identification.
  • Thermoplastic elastomer construction, hydrocarbon-resistant formulation.
  • Maximum working pressure approximately 35 psi at 73°F for this nominal size.
  • Shore A hardness around 57.
  • Operating temperature range -35°F to 165°F (-37°C to 74°C).
  • Typical applications include fuel and lubricant transfer, small engine fuel lines, and petroleum/alcohol blends.
Catalog No. 50-236-5778 Supplier Accuflow Systems Supplier No. AAG00012
Flexible fuel and lubricant tubing formulated to resist swelling and hardening from hydrocarbon-based fluids. Suitable for small-engine fuel lines and general fuel and lubricant transfer applications.
  • 3/16 in inner diameter, 5/16 in outer diameter.
  • 1/16 in wall thickness.
  • 50 ft length, bulk coil.
  • Translucent yellow for fluid identification.
  • Thermoplastic elastomer construction, hydrocarbon-resistant formulation.
  • Maximum working pressure approximately 35 psi at 73°F for this nominal size.
  • Shore A hardness around 57.
  • Operating temperature range -35°F to 165°F (-37°C to 74°C).
  • Typical applications include fuel and lubricant transfer, small engine fuel lines, and petroleum/alcohol blends.