Learn More
Accuflow Systems Saint-Gobain Tygon E-3603 Tubing, 3/16 in ID, 1/4 in OD
Catalog No. 502365784
Encompass_Preferred
Tygon E-3603 thermoplastic tubing is designed for laboratory fluid transfer, peristaltic pump use, and light vacuum service. The phthalate-free formulation provides flexible, thin-walled tubing with consistent dimensional stability and low flow resistance. Packaged in a 50 ft coil for bench and instrument applications.
  • 3/16 in inner diameter, 1/4 in outer diameter, 1/32 in wall
  • 50 ft (15 m) packaged length
  • Phthalate-free formulation
  • Minimum bend radius 1 in (25.4 mm)
  • Maximum working pressure 14 psi (1.0 bar) at 73°F
  • Vacuum rating 9 inHg (229 mmHg)
  • Flexible thin wall for low flow resistance
Catalog No. 50-236-5784 Supplier Accuflow Systems Supplier No. ACF00011
Tygon E-3603 thermoplastic tubing is designed for laboratory fluid transfer, peristaltic pump use, and light vacuum service. The phthalate-free formulation provides flexible, thin-walled tubing with consistent dimensional stability and low flow resistance. Packaged in a 50 ft coil for bench and instrument applications.
  • 3/16 in inner diameter, 1/4 in outer diameter, 1/32 in wall
  • 50 ft (15 m) packaged length
  • Phthalate-free formulation
  • Minimum bend radius 1 in (25.4 mm)
  • Maximum working pressure 14 psi (1.0 bar) at 73°F
  • Vacuum rating 9 inHg (229 mmHg)
  • Flexible thin wall for low flow resistance