Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, 3/16 in ID, 50 ft
Catalog No. 502365880
Encompass_Preferred
Tygon E-3603 tubing is a crystal-clear, flexible thermoplastic tubing designed for reliable fluid transfer and visual inspection in medical, diagnostic, and laboratory instruments. Its high dimensional tolerance and resistance to flex-fatigue support consistent flow and minimize carryover.
  • Crystal-clear, flexible thermoplastic tubing suitable for laboratory and medical applications.
  • Inner diameter 3/16 in, outer diameter 5/16 in, wall thickness 1/16 in.
  • 50 ft package length for extended runs and installations.
  • High dimensional tolerance and resistance to flex-fatigue for consistent fluid transfer.
  • Glassy-smooth inner bore reduces buildup and carryover.
  • BPA- and phthalate-free, low extractables formulation.
Catalog No. 50-236-5880 Supplier Accuflow Systems Supplier No. ACF00012
Tygon E-3603 tubing is a crystal-clear, flexible thermoplastic tubing designed for reliable fluid transfer and visual inspection in medical, diagnostic, and laboratory instruments. Its high dimensional tolerance and resistance to flex-fatigue support consistent flow and minimize carryover.
  • Crystal-clear, flexible thermoplastic tubing suitable for laboratory and medical applications.
  • Inner diameter 3/16 in, outer diameter 5/16 in, wall thickness 1/16 in.
  • 50 ft package length for extended runs and installations.
  • High dimensional tolerance and resistance to flex-fatigue for consistent fluid transfer.
  • Glassy-smooth inner bore reduces buildup and carryover.
  • BPA- and phthalate-free, low extractables formulation.