Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, 3/8 in ID, 50 ft
Catalog No. 502365799
Encompass_Preferred
Saint-Gobain Tygon E-3603 tubing is a clear, flexible thermoplastic tubing engineered for medical, diagnostic, and laboratory fluid transfer. It provides high clarity, low extractables, and resistance to flex-fatigue and abrasion, with a smooth inner bore that minimizes buildup and carryover for reliable, reproducible fluid handling. The 3/8 in inside diameter, 1/2 in outside diameter, and 1/16 in wall thickness are supplied on a 50 ft coil length suitable for bench-scale or instrument installation.
  • Clear, flexible thermoplastic construction for visual inspection.
  • 3/8 in inside diameter, 1/2 in outside diameter, 1/16 in wall thickness.
  • 50 ft (15 m) coil length for instrument runs or bench use.
  • Phthalate-free, low extractables formulation to protect sensitive reagents.
  • Smooth inner bore reduces buildup and carryover.
  • Resistant to flex-fatigue and abrasion for extended service life.
Catalog No. 50-236-5799 Supplier Accuflow Systems Supplier No. ACF00027F
Saint-Gobain Tygon E-3603 tubing is a clear, flexible thermoplastic tubing engineered for medical, diagnostic, and laboratory fluid transfer. It provides high clarity, low extractables, and resistance to flex-fatigue and abrasion, with a smooth inner bore that minimizes buildup and carryover for reliable, reproducible fluid handling. The 3/8 in inside diameter, 1/2 in outside diameter, and 1/16 in wall thickness are supplied on a 50 ft coil length suitable for bench-scale or instrument installation.
  • Clear, flexible thermoplastic construction for visual inspection.
  • 3/8 in inside diameter, 1/2 in outside diameter, 1/16 in wall thickness.
  • 50 ft (15 m) coil length for instrument runs or bench use.
  • Phthalate-free, low extractables formulation to protect sensitive reagents.
  • Smooth inner bore reduces buildup and carryover.
  • Resistant to flex-fatigue and abrasion for extended service life.