Learn More
Accuflow Systems Saint-Gobain Tygon E-3603 Tubing, Special PVC, 3/8 In, 15 m
Catalog No. 502365902
Encompass_Preferred
Clear special PVC process tubing for laboratory and process fluid transfer. The tubing combines flexibility, low permeability, and a glassy smooth inner bore for easy cleaning; coils are marked for measuring and the tubing holds full vacuum at room temperature.
  • 3/8 in inner diameter, 9/16 in outer diameter, 3/32 in wall.
  • 15 m length per coil.
  • Special PVC material, clear.
  • Operating temperature range -46°C to +74°C.
  • Maximum working pressure 1.4 bar at 23°C.
  • Meets FDA, EU food, NSF 51, USP Class VI, and Japan food standards; Prop 65 noted.
  • Does not contain BPA or phthalates.
  • Not peristaltic pump compatible.
Catalog No. 50-236-5902 Supplier Accuflow Systems Supplier No. ACF000028
Clear special PVC process tubing for laboratory and process fluid transfer. The tubing combines flexibility, low permeability, and a glassy smooth inner bore for easy cleaning; coils are marked for measuring and the tubing holds full vacuum at room temperature.
  • 3/8 in inner diameter, 9/16 in outer diameter, 3/32 in wall.
  • 15 m length per coil.
  • Special PVC material, clear.
  • Operating temperature range -46°C to +74°C.
  • Maximum working pressure 1.4 bar at 23°C.
  • Meets FDA, EU food, NSF 51, USP Class VI, and Japan food standards; Prop 65 noted.
  • Does not contain BPA or phthalates.
  • Not peristaltic pump compatible.