Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, Thermoplastic, 5/16 in ID, 50 ft
Catalog No. NC2587857
Encompass_Preferred
Saint-Gobain Tygon E-3603 tubing is a clear, flexible thermoplastic tubing designed for medical, diagnostic, and laboratory fluid handling where chemical compatibility and low extractables are required. It provides visual flow inspection, chemical resistance to many inorganic reagents, and an extra-heavy wall construction for vacuum and pressure applications.
  • Clear thermoplastic construction for visual inspection and low extractables.
  • Nominal inside diameter 5/16 in (7.9 mm), outside diameter 1/2 in (12.7 mm), wall thickness 3/32 in (2.4 mm), length 50 ft (15 m).
  • Maximum recommended operating temperature 165 °F (74 °C); brittle temperature -51 °F (-46 °C).
  • Maximum working pressure at 73 °F: 23 psi (1.6 bar); vacuum rating 29.9 inHg (760 mmHg).
  • BPA-free and phthalate-free formulation to minimize extractables.
  • Extra-heavy wall construction for improved durability and vacuum resistance.
Catalog No. NC2587857 Supplier Accuflow Systems Supplier No. ACF00023
Saint-Gobain Tygon E-3603 tubing is a clear, flexible thermoplastic tubing designed for medical, diagnostic, and laboratory fluid handling where chemical compatibility and low extractables are required. It provides visual flow inspection, chemical resistance to many inorganic reagents, and an extra-heavy wall construction for vacuum and pressure applications.
  • Clear thermoplastic construction for visual inspection and low extractables.
  • Nominal inside diameter 5/16 in (7.9 mm), outside diameter 1/2 in (12.7 mm), wall thickness 3/32 in (2.4 mm), length 50 ft (15 m).
  • Maximum recommended operating temperature 165 °F (74 °C); brittle temperature -51 °F (-46 °C).
  • Maximum working pressure at 73 °F: 23 psi (1.6 bar); vacuum rating 29.9 inHg (760 mmHg).
  • BPA-free and phthalate-free formulation to minimize extractables.
  • Extra-heavy wall construction for improved durability and vacuum resistance.