Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, E-3603, 5/8 in I.D., 50 ft
Catalog No. 502365807
Encompass_Preferred
Tygon E-3603 tubing is a crystal-clear, flexible PVC tubing formulated for phthalate-free medical and laboratory fluid transfer. With a 5/8 in inner diameter, 13/16 in outer diameter, and 3/32 in wall thickness, the 50 ft length provides consistent performance for diagnostic analyzers, IVD systems, and general medical instrumentation.
  • Crystal-clear, flexible PVC tubing for medical and laboratory use.
  • 5/8 in inner diameter and 13/16 in outer diameter for consistent fluid flow.
  • 3/32 in wall thickness for durability and kink resistance.
  • 50 ft package length for extended runs and reduced joins.
  • Phthalate-free formulation suitable for non-DEHP applications.
  • Suitable for diagnostic analyzers, IVD systems, and general medical instrumentation.
Catalog No. 50-236-5807 Supplier Accuflow Systems Supplier No. ACF00045
Tygon E-3603 tubing is a crystal-clear, flexible PVC tubing formulated for phthalate-free medical and laboratory fluid transfer. With a 5/8 in inner diameter, 13/16 in outer diameter, and 3/32 in wall thickness, the 50 ft length provides consistent performance for diagnostic analyzers, IVD systems, and general medical instrumentation.
  • Crystal-clear, flexible PVC tubing for medical and laboratory use.
  • 5/8 in inner diameter and 13/16 in outer diameter for consistent fluid flow.
  • 3/32 in wall thickness for durability and kink resistance.
  • 50 ft package length for extended runs and reduced joins.
  • Phthalate-free formulation suitable for non-DEHP applications.
  • Suitable for diagnostic analyzers, IVD systems, and general medical instrumentation.