Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, E-3603, 7/16 In ID, 9/16 In OD, 50 Ft
Catalog No. 502365916
Encompass_Preferred
Saint-Gobain Tygon E-3603 tubing is a crystal-clear, flexible thermoplastic tubing engineered for medical, diagnostic, and laboratory fluid-transfer applications. It combines high dimensional tolerance, resistance to flex-fatigue, and a glassy-smooth inner bore to minimize buildup and carryover, while its non-oxidizing, non-contaminating formulation reduces extractables for reliable reagent handling.
  • Crystal-clear thermoplastic construction for visual inspection
  • 7/16 in (11.1 mm) inside diameter, 9/16 in (14.3 mm) outside diameter
  • 1/16 in (1.6 mm) wall thickness, 50 ft (15 m) length
  • BPA- and phthalate-free, biocompatible, non-toxic
  • Glassy-smooth inner bore resists buildup and reduces carryover
  • High dimensional tolerance and resistance to flex-fatigue for reproducible fluid transfer
  • Non-oxidizing, non-contaminating formulation minimizes extractables
Catalog No. 50-236-5916 Supplier Accuflow Systems Supplier No. ACF00032
Saint-Gobain Tygon E-3603 tubing is a crystal-clear, flexible thermoplastic tubing engineered for medical, diagnostic, and laboratory fluid-transfer applications. It combines high dimensional tolerance, resistance to flex-fatigue, and a glassy-smooth inner bore to minimize buildup and carryover, while its non-oxidizing, non-contaminating formulation reduces extractables for reliable reagent handling.
  • Crystal-clear thermoplastic construction for visual inspection
  • 7/16 in (11.1 mm) inside diameter, 9/16 in (14.3 mm) outside diameter
  • 1/16 in (1.6 mm) wall thickness, 50 ft (15 m) length
  • BPA- and phthalate-free, biocompatible, non-toxic
  • Glassy-smooth inner bore resists buildup and reduces carryover
  • High dimensional tolerance and resistance to flex-fatigue for reproducible fluid transfer
  • Non-oxidizing, non-contaminating formulation minimizes extractables