Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Tygon ND-100-65, Non-DEHP, 1/4"" ID x 3/8"" OD; 50 ft
Catalog No. NC0911991
Encompass
Formulation: ND-100-65 Applications: Pharmaceutical and biotech research applications. Characteristics:Non-DEHP, phthalate free. Non-wetted surface permits complete drainage. Bondable, weldable. Clear. Certification: USP Class VI, REACH, ISO 10993 Temperature range: -44 to 165¿F (-42 to 74¿C) Sterilize: Autoclave, ethylene oxide, or irradiation.
Catalog No. NC0911991 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9566614
Formulation: ND-100-65 Applications: Pharmaceutical and biotech research applications. Characteristics:Non-DEHP, phthalate free. Non-wetted surface permits complete drainage. Bondable, weldable. Clear. Certification: USP Class VI, REACH, ISO 10993 Temperature range: -44 to 165¿F (-42 to 74¿C) Sterilize: Autoclave, ethylene oxide, or irradiation.