Learn More
AVANTOR MASTERFLEX (VWR LLC) Tygon ultra-soft tubing, 1/8 x 1/4"", 50 feet/pack
Catalog No. NC1685656
Encompass
Formulation: Tygon E-1000 Applications: General laboratory, vent and drain lines. Ideal for lab installations requiring multiple directional changes, abrupt curves, or twisting. Characteristics: High degree of flexibility, performs well at low temperatures, broad chemical resistance, resists kinking and collapse. Transparent. Certification: FDA compliant. Made according to GMP. Temperature range: –67 to 125¿F (–55 to 52¿C) Sterilize:Autoclave or ethylene oxide.
Catalog No. NC1685656 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX6510202
Formulation: Tygon E-1000 Applications: General laboratory, vent and drain lines. Ideal for lab installations requiring multiple directional changes, abrupt curves, or twisting. Characteristics: High degree of flexibility, performs well at low temperatures, broad chemical resistance, resists kinking and collapse. Transparent. Certification: FDA compliant. Made according to GMP. Temperature range: –67 to 125¿F (–55 to 52¿C) Sterilize:Autoclave or ethylene oxide.