Learn More
US Plastic Corporation TYGON 2375 1/4 X 3/8 (10')
SDP
Catalog No. NC1753397
Encompass
Item is sold as 1' each and must be purchased in multiples of 50 per supplier requirements.
1/4"id x 3/8"od x 1/16" wall Tygon 2375 ultra chemical resistant tubing
Catalog No. NC1753397 Supplier US Plastic Corporation Supplier No. 5755050FT
Item is sold as 1' each and must be purchased in multiples of 50 per supplier requirements.
1/4"id x 3/8"od x 1/16" wall Tygon 2375 ultra chemical resistant tubing